Dimethyl-3-oxoglutarate ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate
A0056-25G
A0056-500G
A0056-100G
TCIAA0056-25GEA
470
SEK
TCIAA0056-25G
TCIAA0056-500G
TCIAA0056-100G
Dimethyl-3-oxoglutarate ≥95.0%
Dimethyl-3-oxoglutarate
Formel:
C₇H₁₀O₅ MW: 174,15 g/mol Kokpunkt: 150°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00008462 CAS nummer: 1830-54-2 EINECS: 217-385-8 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...