Cyanoacetamide ≥98.0%

Leverantör: TCI
Varning

C0437-25G C0437-500G
TCIAC0437-25GEA 215 SEK
TCIAC0437-25G TCIAC0437-500G
Cyanoacetamide ≥98.0%
Cyanoacetamide
Formel: C₃H₄N₂O
MDL Number: MFCD00008024
CAS nummer: 107-91-5
EINECS: 203-531-8
Merck Index: 14,02688
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f976977b0f077437785870912bbbc2b110312e0e&vid=f976977b0f077437785870912bbbc2b110312e0e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 122.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR