(±)-Epoxystyrene 98,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

Synonyms: Styrene oxide

E0013-25ML E0013-500ML
TCIAE0013-25MLEA 230 SEK
TCIAE0013-25ML TCIAE0013-500ML
(±)-Epoxystyrene 98,0 GC_ANALYSMETOD
(±)-Epoxystyrene
Formel: C₈H₈O
MW: 120.15 g/mol
Kokpunkt: 194°C
Smältpunkt: –37°C
Flampunkt: 77°C
MDL Number: MFCD00005121
CAS nummer: 96-09-3
EINECS: 202-476-7
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecde058eb1ba07c20107aa092d6ea18901b6feb0&vid=ecde058eb1ba07c20107aa092d6ea18901b6feb0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR