3-Quinuclidinone hydrochloride ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

Q0052-25G Q0052-5G
TCIAQ0052-25GEA 1025 SEK
TCIAQ0052-25G TCIAQ0052-5G
3-Quinuclidinone hydrochloride ≥98.0%
3-Quinuclidinone hydrochloride
Formel: C₇H₁₂ClNO
MDL Number: MFCD00137391
CAS nummer: 1193-65-3
EINECS: 214-776-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2822e60c67a97dace89fd239cda9c7b0e79a71d&vid=c2822e60c67a97dace89fd239cda9c7b0e79a71d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR