Furfurylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 2-(Aminomethyl)furan

F0091-25ML F0091-500ML F0091-100ML
TCIAF0091-25MLEA 230 SEK
TCIAF0091-25ML TCIAF0091-500ML TCIAF0091-100ML
Furfurylamine ≥98.0% (by GC, titration analysis)
Furfurylamine
Formel: C₅H₇NO
MW: 97,12 g/mol
Kokpunkt: 146°C
Flampunkt: 37°C
MDL Number: MFCD00003258
CAS nummer: 617-89-0
EINECS: 210-536-9
UN: 2526
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cb3bc2bf6048aaea7d989f860334b1079d8af64&vid=3cb3bc2bf6048aaea7d989f860334b1079d8af64&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR