1,3-Dibromo-5,5-dimethylhydantoin ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1,3-Dibromo-5,5-dimethylimidazolidine-2,4-dione
D1265-25G
D1265-500G
TCIAD1265-25GEA
215
SEK
TCIAD1265-25G
TCIAD1265-500G
1,3-Dibromo-5,5-dimethylhydantoin ≥97.0%
1,3-Dibromo-5,5-dimethylhydantoin
Formel:
C₅H₆Br₂N₂O₂ MW: 285,92 g/mol |
MDL Number:
MFCD00003189 CAS nummer: 77-48-5 EINECS: 201-030-9 UN: 3087 ADR: 5.1,II Merck Index: 14,03017 |
Specifikation testresultat
Appearance | White to Light red to Green powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Solubility in hot Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...