Hydantoin ≥99.0%

Leverantör: TCI EUROPE

Synonyms: Imidazolidine-2,4-dione

H0167-500G H0167-25G
TCIAH0167-500GEA 965 SEK
TCIAH0167-500G TCIAH0167-25G
Hydantoin ≥99.0%
Hydantoin
Formel: C₃H₄N₂O₂
MW: 100,08 g/mol
MDL Number: MFCD00005259
CAS nummer: 461-72-3
EINECS: 207-313-3
Merck Index: 14,04760
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=980d9e83235c7897299e4fb1f6c3c53823e9d3f2&vid=980d9e83235c7897299e4fb1f6c3c53823e9d3f2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 221.0 to 225.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR