(±)-β-Aminophenethyl alcohol ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol

P1792-25G P1792-5G
TCIAP1792-25GEA 3375 SEK
TCIAP1792-25G TCIAP1792-5G
(±)-β-Aminophenethyl alcohol ≥98.0% (by GC, titration analysis)
(±)-β-Aminophenethyl alcohol
Formel: C₈H₁₁NO
Kokpunkt: 106°C
Storage Temperature: Kylskåp
MDL Number: MFCD00130145
CAS nummer: 7568-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0caa21b1dbb6084565800091a49990102c15f0da&vid=0caa21b1dbb6084565800091a49990102c15f0da&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR