6-Aminopicolinic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Aminopyridine-2-carboxylic acid
A2188-1G
A2188-5G
TCIAA2188-1GEA
470
SEK
TCIAA2188-1G
TCIAA2188-5G
6-Aminopicolinic acid ≥98.0% (by HPLC, titration analysis)
6-Aminopicolinic acid
Formel:
C₆H₆N₂O₂ MW: 138,13 g/mol Smältpunkt: 315°C |
MDL Number:
MFCD00233711 CAS nummer: 23628-31-1 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...