Tributyl phosphite ≥93.0%

Leverantör: TCI
Varning

Synonyms: Tri-n-butylphosphite

T0362-25ML T0362-500ML
TCIAT0362-25MLEA 215 SEK
TCIAT0362-25ML TCIAT0362-500ML
Tributyl phosphite ≥93.0%
Tributyl phosphite
Formel: C₁₂H₂₇O₃P
MW: 250.32 g/mol
Flampunkt: 129°C
MDL Number: MFCD00009437
CAS nummer: 102-85-2
EINECS: 203-061-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d222230e1915e9ea51cfe511ca8f2fa1485542f1&vid=d222230e1915e9ea51cfe511ca8f2fa1485542f1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR