N-Methylphenethylamine ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
M1950-5G
M1950-25G
TCIAM1950-5GEA
845
SEK
TCIAM1950-5G
TCIAM1950-25G
N-Methylphenethylamine ≥97.0% (by GC, titration analysis)
N-Methylphenethylamine
Formel:
C₉H₁₃N MW: 135,21 g/mol Kokpunkt: 205°C Flampunkt: 73°C |
MDL Number:
MFCD00008291 CAS nummer: 589-08-2 EINECS: 209-632-3 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...