N-tert-Butylbenzylamine ≥96.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N-Benzyl-tert-butylamine, N-Benzyl-2-methylpropan-2-amine

B1176-25ML B1176-500ML
TCIAB1176-25MLEA 455 SEK
TCIAB1176-25ML TCIAB1176-500ML
N-tert-Butylbenzylamine ≥96.0%
N-tert-Butylbenzylamine
Formel: C₁₁H₁₇N
MW: 163,26 g/mol
Kokpunkt: 252°C
Flampunkt: 90°C
MDL Number: MFCD00008798
CAS nummer: 3378-72-1
EINECS: 222-179-6
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ca9cd1bcf24a434f2e352345d87a4732c79a1f9&vid=7ca9cd1bcf24a434f2e352345d87a4732c79a1f9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR