2'-Methylformanilide ≥98.0%

Leverantör: TCI EUROPE
Varning

M1431-25G M1431-5G
TCIAM1431-25GEA 935 SEK
TCIAM1431-25G TCIAM1431-5G
2'-Methylformanilide ≥98.0%
2'-Methylformanilide
Formel: C₈H₉NO
MW: 135,17 g/mol
MDL Number: MFCD00014122
CAS nummer: 94-69-9
EINECS: 202-355-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c26b168bb46641bf599b8d4e1a238c3edc2c9ce8&vid=c26b168bb46641bf599b8d4e1a238c3edc2c9ce8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR