Succinyl chloride ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Succinyl dichloride

S0110-25G S0110-250G
TCIAS0110-25GEA 415 SEK
TCIAS0110-25G TCIAS0110-250G
Succinyl chloride ≥95.0%
Succinyl chloride
Formel: C₄H₄Cl₂O₂
MW: 154,98 g/mol
Kokpunkt: 192°C
Flampunkt: 76°C
MDL Number: MFCD00000749
CAS nummer: 543-20-4
EINECS: 208-838-0
UN: 3265
ADR: 8,III
Merck Index: 14,08873
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c12aacbcd32fa7bb574639fe1d1e7b4be3e6f3e6&vid=c12aacbcd32fa7bb574639fe1d1e7b4be3e6f3e6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR