Methoxyacetonitrile ≥98.0%
Leverantör: TCI
|
Varning
|
M0103-25ML
M0103-5ML
TCIAM0103-25MLEA
2200
SEK
TCIAM0103-25ML
TCIAM0103-5ML
Methoxyacetonitrile ≥98.0%
Methoxyacetonitrile
Formel:
C₃H₅NO Kokpunkt: 119°C Flampunkt: 32°C |
MDL Number:
MFCD00001892 CAS nummer: 1738-36-9 EINECS: 217-092-5 UN: 1993 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...