Dichlorodiethylsilane ≥98.0%
Leverantör: TCI
|
Fara
|
D1976-25G
D1976-5G
TCIAD1976-25GEA
4575
SEK
TCIAD1976-25G
TCIAD1976-5G
Dichlorodiethylsilane ≥98.0%
Dichlorodiethylsilane
Formel:
C₄H₁₀Cl₂Si Kokpunkt: 129°C Smältpunkt: -97°C Flampunkt: 24°C |
MDL Number:
MFCD00000493 CAS nummer: 1719-53-5 EINECS: 217-005-0 UN: 1767 ADR: 8,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...