1-Methylcyclohexene ≥98.0%

Leverantör: TCI
Fara

M0200-25ML M0200-500ML M0200-100ML
TCIAM0200-25MLEA 735 SEK
TCIAM0200-25ML TCIAM0200-500ML TCIAM0200-100ML
1-Methylcyclohexene ≥98.0%
1-Methylcyclohexene
Formel: C₇H₁₂
Kokpunkt: 110°C
Densitet: 0,8102 g/cm³ (20 °C)
Flampunkt: -3°C
Storage Temperature: Omgivande
MDL Number: MFCD00001548
CAS nummer: 591-49-1
EINECS: 209-718-0
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d06c63674ecb9f04641ee412311de8b74b2368a&vid=5d06c63674ecb9f04641ee412311de8b74b2368a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR