cis-1,4-Dimetylcyklohexan ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Cis-1,4-dimethylcyclohexane
D1715-25ML
D1715-5ML
TCIAD1715-25MLEA
1925
SEK
TCIAD1715-25ML
TCIAD1715-5ML
cis-1,4-Dimetylcyklohexan ≥98.0%
cis-1,4-Dimetylcyklohexan
Formel:
C₈H₁₆ MW: 112,22 g/mol Kokpunkt: 125°C Smältpunkt: –42 °C Densitet: 1,283 g/cm³ (20 °C) Flampunkt: 6°C Storage Temperature: Omgivande |
MDL Number:
MFCD00064174 CAS nummer: 624-29-3 EINECS: 210-840-1 UN: 2263 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...