Methylcyclopentane ≥96.0%

Leverantör: TCI EUROPE
Fara

M0203-25ML M0203-500ML
TCIAM0203-25MLEA 230 SEK
TCIAM0203-25ML TCIAM0203-500ML
Methylcyclopentane ≥96.0%
Methylcyclopentane
Formel: C₆H₁₂
MW: 84,16 g/mol
Kokpunkt: 72°C
Smältpunkt: –142 °C
Densitet: 0,74 g/cm³ (20 °C)
Flampunkt: -29°C
Storage Temperature: Omgivande
MDL Number: MFCD00001382
CAS nummer: 96-37-7
EINECS: 202-503-2
UN: 2298
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9556970008ca42a6ee0e18e1b42e21db129dbc6f&vid=9556970008ca42a6ee0e18e1b42e21db129dbc6f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR