Ethylcyclohexane ≥99.0%

Leverantör: TCI EUROPE
Fara

E0073-25ML E0073-500ML
TCIAE0073-25MLEA 215 SEK
TCIAE0073-25ML TCIAE0073-500ML
Ethylcyclohexane ≥99.0%
Ethylcyclohexane
Formel: C₈H₁₆
MW: 112,22 g/mol
Kokpunkt: 132°C
Smältpunkt: –111,1 °C
Densitet: 0,788 g/cm³ (20 °C)
Flampunkt: 16°C
Storage Temperature: Omgivande
MDL Number: MFCD00001523
CAS nummer: 1678-91-7
EINECS: 216-835-0
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=068ae1fc8211cc4c69222f78b5faab17d9238e97&vid=068ae1fc8211cc4c69222f78b5faab17d9238e97&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR