Butylcyclohexane 99,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
B0822-25ML
B0822-500ML
TCIAB0822-25MLEA
485
SEK
TCIAB0822-25ML
TCIAB0822-500ML
Butylcyclohexane 99,0 GC_ANALYSMETOD
Butylcyclohexane
Formel:
C₁₀H₂₀ MW: 140.27 g/mol Kokpunkt: 180°C Smältpunkt: -78 Flampunkt: 41°C Storage Temperature: Omgivande |
MDL Number:
MFCD00001530 CAS nummer: 1678-93-9 EINECS: 216-837-1 UN: 3295 ADR: 3 III |
|
Specifikation testresultat
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...