Isopropylcyclohexane ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Hexahydrocumene

I0161-25ML I0161-500ML
TCIAI0161-25MLEA 310 SEK
TCIAI0161-25ML TCIAI0161-500ML
Isopropylcyclohexane ≥99.0%
Isopropylcyclohexane
Formel: C₉H₁₈
MW: 126,24 g/mol
Kokpunkt: 151°C
Smältpunkt: –90 °C
Densitet: 0,8036 g/cm³ (20 °C)
Flampunkt: 35°C
Storage Temperature: Omgivande
MDL Number: MFCD00001480
CAS nummer: 696-29-7
EINECS: 211-792-4
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70e10e5249caabaf28e31f066c1bdab7ee4b246b&vid=70e10e5249caabaf28e31f066c1bdab7ee4b246b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR