(Chloromethyl)cyclopropane ≥95.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Cyclopropyl methyl chloride
C2469-25G C2469-5G
TCIAC2469-25GEA 1275 SEK
TCIAC2469-25G TCIAC2469-5G
(Chloromethyl)cyclopropane ≥95.0%
(Chloromethyl)cyclopropane
Formel: C₄H₇Cl
MW: 90,55 g/mol
Kokpunkt: 88°C
Smältpunkt: –91°C
Densitet: 0,98 g/cm³ (20 °C)
Flampunkt: –2°C
Storage Temperature: Omgivande
MDL Number: MFCD00001307
CAS nummer: 5911-08-0
EINECS: 227-632-1
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d8706ca3f338fce9d9781df29930822f8a52ebf&vid=2d8706ca3f338fce9d9781df29930822f8a52ebf&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %