4-Metylcyklohexanon ≥98.0%

Leverantör: TCI EUROPE
Varning

M0199-25ML M0199-500ML
TCIAM0199-25MLEA 255 SEK
TCIAM0199-25ML TCIAM0199-500ML
4-Metylcyklohexanon ≥98.0%
4-Metylcyklohexanon
Formel: C₇H₁₂O
MW: 112,17 g/mol
Kokpunkt: 171°C
Smältpunkt: –41 °C
Densitet: 0,92 g/cm³ (20 °C)
Flampunkt: 48°C
Storage Temperature: Omgivande
MDL Number: MFCD00001643
CAS nummer: 589-92-4
EINECS: 209-665-3
UN: 2297
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=787255fa887b127c5bbfb3732b665c9bf69127de&vid=787255fa887b127c5bbfb3732b665c9bf69127de&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR