2-Acetamido-5-bromopyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Acetylamino-5-bromopyridine, N-(5-Bromopyridin-2-yl)acetamide

A2375-1G A2375-5G
TCIAA2375-1GEA 215 SEK
TCIAA2375-1G TCIAA2375-5G
2-Acetamido-5-bromopyridine ≥98.0%
2-Acetamido-5-bromopyridine
Formel: C₇H₇BrN₂O
MW: 215,05 g/mol
MDL Number: MFCD00468968
CAS nummer: 7169-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a599342ad8b489454cdc4b7eaa59fd7c882b909a&vid=a599342ad8b489454cdc4b7eaa59fd7c882b909a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR