Ethyl-5-bromo-2-chlorobenzoate ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Ethyl 5-bromo-2-chlorobenzoate
B2255-25G B2255-5G
TCIAB2255-25GEA 1525 SEK
TCIAB2255-25G TCIAB2255-5G
Ethyl-5-bromo-2-chlorobenzoate ≥98.0%
Ethyl-5-bromo-2-chlorobenzoate
Formel: C₉H₈BrClO₂
MW: 263,52 g/mol
MDL Number: MFCD00013635
CAS nummer: 76008-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c92ebab969f0540c0dc012b07b5efe351e866d6&vid=1c92ebab969f0540c0dc012b07b5efe351e866d6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %