L-α-Aminoadipic acid ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid
A1457-1G
A1457-5G
TCIAA1457-1GEA
1525
SEK
TCIAA1457-1G
TCIAA1457-5G
L-α-Aminoadipic acid ≥97.0% (by GC, titration analysis)
L-α-Aminoadipic acid
Formel:
C₆H₁₁NO₄ MW: 161,16 g/mol |
MDL Number:
MFCD00002636 CAS nummer: 1118-90-7 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Specific rotation [a]20/D | +23.0 to +26.0 deg (C=5, 6mol/L HCl) |
Solubility in HCl (1+1) | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...