Bis(2-methoxyethyl)adipate ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Adipic acid bis(2-methoxyethyl) ester
A0165-25ML A0165-500ML
TCIAA0165-25MLEA 815 SEK
TCIAA0165-25ML TCIAA0165-500ML
Bis(2-methoxyethyl)adipate ≥98.0%
Bis(2-methoxyethyl)adipate
Formel: C₁₂H₂₂O₆
MW: 262,3 g/mol
Kokpunkt: 205°C
MDL Number: MFCD00041905
CAS nummer: 106-00-3
EINECS: 203-352-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e8f38fdca1b9635ca7f32cf6ee19ce1f54cacf7&vid=4e8f38fdca1b9635ca7f32cf6ee19ce1f54cacf7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %