1-Butylpyridiniumtetrafluoroborate ≥94.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Butylpyridin-1-ium tetrafluoroborate

B3232-25G B3232-5G
TCIAB3232-25GEA 2275 SEK
TCIAB3232-25G TCIAB3232-5G
1-Butylpyridiniumtetrafluoroborate ≥94.0%
1-Butylpyridiniumtetrafluoroborate
Formel: C₉H₁₄BF₄N
MW: 223,02 g/mol
MDL Number: MFCD03427602
CAS nummer: 203389-28-0
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f238bf4e09b2fee478a75a6981d75ca90637b197&vid=f238bf4e09b2fee478a75a6981d75ca90637b197&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR