Methyl-2-furoate ≥99.0%

Leverantör: TCI
Fara

F0075-25ML F0075-250ML
TCIAF0075-25MLEA 295 SEK
TCIAF0075-25ML TCIAF0075-250ML
Methyl-2-furoate ≥99.0%
Methyl-2-furoate
Formel: C₆H₆O₃
Flampunkt: 73°C
MDL Number: MFCD00003236
CAS nummer: 611-13-2
EINECS: 210-254-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5080b7b606152c8702a0adf2c87e78cf0f56e0ec&vid=5080b7b606152c8702a0adf2c87e78cf0f56e0ec&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR