2-Furoyl chloride ≥98.0%
Leverantör: TCI
|
Fara
|
F0233-25G
F0233-500G
TCIAF0233-25GEA
295
SEK
TCIAF0233-25G
TCIAF0233-500G
2-Furoyl chloride ≥98.0%
2-Furoyl chloride
Formel:
C₅H₃ClO₂ Flampunkt: 85°C |
MDL Number:
MFCD00003228 CAS nummer: 527-69-5 EINECS: 208-422-9 UN: 3265 ADR: 8,II Merck Index: 14,04310 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...