4-Picolyl chloride hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride

C0805-25G C0805-250G C0805-5G
TCIAC0805-25GEA 975 SEK
TCIAC0805-25G TCIAC0805-250G TCIAC0805-5G
4-Picolyl chloride hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Picolyl chloride hydrochloride
Formel: C₆H₇Cl₂N
MW: 164,03 g/mol
MDL Number: MFCD00012826
CAS nummer: 1822-51-1
EINECS: 217-350-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d49d07c8646c0babb87121d97eab2b52192ba30&vid=4d49d07c8646c0babb87121d97eab2b52192ba30&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR