Piperidine-N-carbaldehyde ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Formylpiperidine, N-Formylpiperidine

F0234-25ML F0234-500ML F0234-100ML
TCIAF0234-25MLEA 215 SEK
TCIAF0234-25ML TCIAF0234-500ML TCIAF0234-100ML
Piperidine-N-carbaldehyde ≥98.0%
Piperidine-N-carbaldehyde
Formel: C₆H₁₁NO
MW: 113,16 g/mol
Kokpunkt: 222°C
Flampunkt: 91°C
MDL Number: MFCD00006483
CAS nummer: 2591-86-8
EINECS: 219-986-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb87216c2c409a29b19fbfe3955b721d17d0186e&vid=bb87216c2c409a29b19fbfe3955b721d17d0186e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR