4-Chloro-2-iodobenzoic acid ≥98.0%
Leverantör: TCI
Varning
C2604-1G C2604-5G
TCIAC2604-1GEA 510 SEK
TCIAC2604-1G TCIAC2604-5G
4-Chloro-2-iodobenzoic acid ≥98.0%
4-Chloro-2-iodobenzoic acid
Formel: C₇H₄ClIO₂
MW: 282.46 g/mol
MDL Number: MFCD00060676
CAS nummer: 13421-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b22a93c2c2cfaa95f8670582722607512f1c351b&vid=b22a93c2c2cfaa95f8670582722607512f1c351b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 162.0 to 166.0 deg-C