2-Bromo-6-methoxypyridine ≥97.0%

Leverantör: TCI EUROPE
Varning

B3260-25G B3260-5G
TCIAB3260-25GEA 1475 SEK
TCIAB3260-25G TCIAB3260-5G
2-Bromo-6-methoxypyridine ≥97.0%
2-Bromo-6-methoxypyridine
Formel: C₆H₆BrNO
MW: 188,02 g/mol
Kokpunkt: 91°C
Flampunkt: 104°C
MDL Number: MFCD00088345
CAS nummer: 40473-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9111fe8d48130382ce3858b9c098367ce6d7534a&vid=9111fe8d48130382ce3858b9c098367ce6d7534a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR