Methyl-3-bromopropionate ≥99.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Methyl 3-bromopropanoate
B0648-25G
B0648-100G
TCIAB0648-25GEA
670
SEK
TCIAB0648-25G
TCIAB0648-100G
Methyl-3-bromopropionate ≥99.0%
Methyl-3-bromopropionate
Formel:
C₄H₇BrO₂ Kokpunkt: 105°C Flampunkt: 75°C |
MDL Number:
MFCD00000250 CAS nummer: 3395-91-3 EINECS: 222-247-5 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...