Cycloheptane ≥98.0%

Leverantör: TCI EUROPE
Fara

C0465-25ML C0465-5ML
TCIAC0465-25MLEA 2000 SEK
TCIAC0465-25ML TCIAC0465-5ML
Cycloheptane ≥98.0%
Cycloheptane
Formel: C₇H₁₄
MW: 98,19 g/mol
Kokpunkt: 118°C
Smältpunkt: –12 °C
Densitet: 0,8093 g/cm³ (20 °C)
Flampunkt: 15°C
Storage Temperature: Omgivande
MDL Number: MFCD00004147
CAS nummer: 291-64-5
EINECS: 206-030-2
UN: 2241
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d4ec041af1d3e66769171f1eabb4c95285ccfdd&vid=3d4ec041af1d3e66769171f1eabb4c95285ccfdd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR