Cyclobutanol ≥98.0%
Leverantör: TCI
|
Fara
|
C0770-1G
C0770-5G
TCIAC0770-1GEA
765
SEK
TCIAC0770-1G
TCIAC0770-5G
Cyclobutanol ≥98.0%
Cyclobutanol
Formel:
C₄H₈O MW: 72.11 g/mol Kokpunkt: 125°C Flampunkt: 21°C Storage Temperature: Omgivande |
MDL Number:
MFCD00001318 CAS nummer: 2919-23-5 EINECS: 220-858-1 UN: 1987 ADR: 3 II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...