Cyclopropylamine ≥95.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: Aminocyclopropane
C0981-25ML C0981-5ML
TCIAC0981-25MLEA 590 SEK
TCIAC0981-25ML TCIAC0981-5ML
Cyclopropylamine ≥95.0% (by GC, titration analysis)
Cyclopropylamine
Formel: C₃H₇N
MW: 57.1 g/mol
Kokpunkt: 50°C
Smältpunkt: -10
Densitet: 0.8131
Flampunkt: 1°C
Storage Temperature: Omgivande
MDL Number: MFCD00001301
CAS nummer: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e619f5aaeb6f86cfa140bf8b3a1f8b211c7549ae&vid=e619f5aaeb6f86cfa140bf8b3a1f8b211c7549ae&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %