Cyclopentylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Fara

C0887-25ML C0887-500ML
TCIAC0887-25MLEA 445 SEK
TCIAC0887-25ML TCIAC0887-500ML
Cyclopentylamine ≥98.0% (by GC, titration analysis)
Cyclopentylamine
Formel: C₅H₁₁N
Kokpunkt: 107°C
Smältpunkt: –86°C
Densitet: 0,8689 g/cm³ (20 °C)
Flampunkt: 13°C
Storage Temperature: Omgivande
MDL Number: MFCD00001380
CAS nummer: 1003-03-8
EINECS: 213-697-3
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b5b6bd6c2ec807c1496052878f62bd58bb97b20&vid=5b5b6bd6c2ec807c1496052878f62bd58bb97b20&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR