Cyclobutylamine ≥97.0% (by GC, titration analysis)
Leverantör: TCI
Fara
A1036-1ML A1036-5ML
TCIAA1036-1MLEA 365 SEK
TCIAA1036-1ML TCIAA1036-5ML
Cyclobutylamine ≥97.0% (by GC, titration analysis)
Cyclobutylamine
Formel: C₄H₉N
Kokpunkt: 82°C
Densitet: 0,8328 g/cm³ (20 °C)
Flampunkt: -1°C
Storage Temperature: Kylskåp
MDL Number: MFCD00001328
CAS nummer: 2516-34-9
EINECS: 219-736-0
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a7ff2475974e8239d03c74d92ef2e854aad6bcb&vid=4a7ff2475974e8239d03c74d92ef2e854aad6bcb&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %