1,1-Cyclobutanedicarboxylic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Cyclobutane-1,1-dicarboxylic acid
C0769-25G C0769-5G
TCIAC0769-25GEA 1275 SEK
TCIAC0769-25G TCIAC0769-5G
1,1-Cyclobutanedicarboxylic acid ≥98.0% (by GC, titration analysis)
1,1-Cyclobutanedicarboxylic acid
Formel: C₆H₈O₄
MW: 144,13 g/mol
Kokpunkt: 663 °C (1013 hPa)
Smältpunkt: 156…158 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001325
CAS nummer: 5445-51-2
EINECS: 226-651-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a42bedf2922e078460da36d25d628f0c37be75f2&vid=a42bedf2922e078460da36d25d628f0c37be75f2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %