1-Aminocyclopentanecarboxylic acid ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Cycloleucine, 1-Aminocyclopentane-1-carboxylic acid
A1063-25G
A1063-5G
A1063-1G
TCIAA1063-25GEA
3325
SEK
TCIAA1063-25G
TCIAA1063-5G
TCIAA1063-1G
1-Aminocyclopentanecarboxylic acid ≥98.0%
1-Aminocyclopentanecarboxylic acid
Formel:
C₆H₁₁NO₂ MW: 129,16 g/mol Kokpunkt: 254 °C (1013 hPa) Smältpunkt: 330°C Storage Temperature: Omgivande |
MDL Number:
MFCD00001381 CAS nummer: 52-52-8 EINECS: 200-144-6 UN: 2811 ADR: 6.1,III Merck Index: 14,02734 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...