1-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 1-Amino-1-cyclohexanecarboxylic acid, 1-Aminocyclohexane-1-carboxylic acid

A1068-100G A1068-25G
TCIAA1068-100GEA 7925 SEK
TCIAA1068-100G TCIAA1068-25G
1-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Aminocyclohexanecarboxylic acid
Formel: C₇H₁₃NO₂
MW: 143,19 g/mol
Kokpunkt: 272 °C (1013 hPa)
Smältpunkt: 170 °C
Densitet: 1,229 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001487
CAS nummer: 2756-85-6
EINECS: 220-411-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7397972b389cc17f098502d2013cf0b8c34530e&vid=14af5f6c8f0da4506c76c49f564ee0daa73f18d8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR