Cyclobutanone ≥97.0% stabilised

Leverantör: TCI EUROPE
Fara


Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

C1913-1G C1913-5G
TCIAC1913-1GEA 485 SEK
TCIAC1913-1G TCIAC1913-5G
Cyclobutanone ≥97.0% stabilised
Cyclobutanone
Formel: C₄H₆O
MW: 70,09 g/mol
Kokpunkt: 99°C
Smältpunkt: -53°C
Densitet: 0,93 g/cm³ (20 °C)
Flampunkt: 10°C
Storage Temperature: Frys
MDL Number: MFCD00001332
CAS nummer: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87df8957053738d3a53d7fd954dc66947bbeeee&vid=fbf2c0aa5ab86f74b1393b8e26f6d8df0ac3b81f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR