6-Methylquinoline ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

M0416-25G M0416-5G
TCIAM0416-25GEA 845 SEK
TCIAM0416-25G TCIAM0416-5G
6-Methylquinoline ≥98.0% (by GC, titration analysis)
6-Methylquinoline
Formel: C₁₀H₉N
MW: 143,19 g/mol
Kokpunkt: 259°C
Smältpunkt: –22,2 °C
Densitet: 1,0815 g/cm³ (0 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006804
CAS nummer: 91-62-3
EINECS: 202-084-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0217bc2ce242f9c6f0d3bb8ef77738b2a18b7f62&vid=0217bc2ce242f9c6f0d3bb8ef77738b2a18b7f62&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR