Quinaldin (2-Metylkinolin) ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
Q0005-25G Q0005-500G Q0005-100G
TCIAQ0005-25GEA 215 SEK
TCIAQ0005-25G TCIAQ0005-500G TCIAQ0005-100G
Quinaldin (2-Metylkinolin) ≥98.0% (by GC, titration analysis)
Quinaldin (2-Metylkinolin)
Formel: C₁₀H₉N
Kokpunkt: 247°C
Smältpunkt: –2°C
Densitet: 1,0585 g/cm³ (20 °C)
Flampunkt: 110°C
Storage Temperature: Omgivande
MDL Number: MFCD00006756
CAS nummer: 91-63-4
EINECS: 202-085-1
Merck Index: 14,08047
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1601dfa5f08e58b01173c87a58719b05b1cc9792&vid=cc30712267a5d72b3c8cd30c61872c447fa99828&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %