2-Chlorolepidine ≥98.0%
Leverantör: TCI
Varning
C0191-25G C0191-5G
TCIAC0191-25GEA 2025 SEK
TCIAC0191-25G TCIAC0191-5G
2-Chlorolepidine ≥98.0%
2-Chlorolepidine
Formel: C₁₀H₈ClN
Kokpunkt: 296°C
Smältpunkt: 55…58 °C
Densitet: 1,23 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006742
CAS nummer: 634-47-9
EINECS: 211-209-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db65ae79d7e76ff3653f5e4618aeed014f1e83e8&vid=47d0bc400aaba8d0b08716a59f63e7e5caa941ab&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 60.0 deg-C