6-Quinolinol ≥96.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Quinolin-6-ol, 6-Hydroxyquinoline
Q0060-25G
Q0060-5G
TCIAQ0060-25GEA
3075
SEK
TCIAQ0060-25G
TCIAQ0060-5G
6-Quinolinol ≥96.0%
6-Quinolinol
Formel:
C₉H₇NO Kokpunkt: 313 °C (1013 hPa) Smältpunkt: 192…193 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00047611 CAS nummer: 580-16-5 EINECS: 209-454-6 |
|
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...