5-Quinolinol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 5-Hydroxyquinoline, Quinolin-5-ol
Q0059-1G Q0059-5G
TCIAQ0059-1GEA 485 SEK
TCIAQ0059-1G TCIAQ0059-5G
5-Quinolinol ≥98.0% (by GC, titration analysis)
5-Quinolinol
Formel: C₉H₇NO
MW: 145,16 g/mol
Kokpunkt: 313 °C (1013 hPa)
Smältpunkt: 223…226 °C
Densitet: 1,57 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006792
CAS nummer: 578-67-6
EINECS: 209-428-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5045dd4cc5ea33f11f796306669504ccc32d7eb&vid=8b92cd77c8d9347962ff15cf146134e8e7ed48cf&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %